3-Methyl-1-(5-nitropyridin-2-yl)piperazine

Supplier: Matrix Scientific

024424-500MG
101818-068EA 375.38 USD
101818-068
3-Methyl-1-(5-nitropyridin-2-yl)piperazine
3-Methyl-1-(5-nitropyridin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₄N₄O₂
MDL Number: MFCD08692454
CAS Number: 773879-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR