4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline

Supplier: Matrix Scientific

Synonyms: 4-((4-Methylpiperazin-1-yl)sulphonyl)aniline, 4-((4-Methylpiperazin-1-yl)sulfonyl)aniline

023878-500MG
101816-956EA 81.54 USD
101816-956
4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline
4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₂S
MDL Number: MFCD03050283
CAS Number: 21623-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR