(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-chloro-4-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

023547-500MG
101816-216EA 315.38 USD
101816-216
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-chloro-4-fluorophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(3-chloro-4-fluorophenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₉ClFNO₅
MDL Number: MFCD08687079
CAS Number: 1354487-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR