3-(4-Fluorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023398-500MG
101815-926EA 427.69 USD
101815-926
3-(4-Fluorophenoxy)pyrrolidine hydrochloride
3-(4-Fluorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClFNO
MW: 217.6700 g/mol
MDL Number: MFCD09879244
CAS Number: 23123-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR