4-(3-Chlorophenoxy)piperidine

Supplier: Matrix Scientific

023283-500MG
101815-700EA 315.38 USD
101815-700
4-(3-Chlorophenoxy)piperidine
4-(3-Chlorophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₄ClNO
MDL Number: MFCD08061087
CAS Number: 97840-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR