4-Chloro-2-fluorophenyl-4-piperidinylmethyl ether

Supplier: Matrix Scientific

023143-500MG
101815-424EA 315.38 USD
101815-424
4-Chloro-2-fluorophenyl-4-piperidinylmethyl ether
4-Chloro-2-fluorophenyl-4-piperidinylmethyl ether

Hazard: Irritant

Formula: C₁₂H₁₅ClFNO
MDL Number: MFCD08687336
CAS Number: 946680-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR