5-Methyl-2-(3-pyridinyloxy)aniline

Supplier: Matrix Scientific

022172-500MG
101813-506EA 315.38 USD
101813-506
5-Methyl-2-(3-pyridinyloxy)aniline
5-Methyl-2-(3-pyridinyloxy)aniline

Hazard: Irritant

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD08688388
CAS Number: 457099-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR