2-(2,3-Dimethylphenoxy)aniline

Supplier: Matrix Scientific

021827-500MG
101812-818EA 315.38 USD
101812-818
2-(2,3-Dimethylphenoxy)aniline
2-(2,3-Dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₅NO
MW: 213.28 g/mol
MDL Number: MFCD07365108
CAS Number: 3169-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR