2-(2-Chlorophenoxy)aniline

Supplier: Matrix Scientific

021820-500MG
101812-804EA 315.38 USD
101812-804
2-(2-Chlorophenoxy)aniline
2-(2-Chlorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₁₀ClNO
MW: 219.67 g/mol
MDL Number: MFCD07365106
CAS Number: 56966-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR