3-Fluoro-3'-methoxy[1,1'-biphenyl]-4-amine

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-3'-methoxy-[1,1'-biphenyl]-4-amine

021709-500MG
101812-592EA 146.15 USD
101812-592
3-Fluoro-3'-methoxy[1,1'-biphenyl]-4-amine
3-Fluoro-3'-methoxy[1,1'-biphenyl]-4-amine

Hazard: Irritant

Formula: C₁₃H₁₂FNO
MDL Number: MFCD06802498
CAS Number: 867287-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR