5-tert-Butyl-2-methoxybenzenesulfonyl chloride

Supplier: Matrix Scientific

021498-500MG
101812-196EA 200 USD
101812-196
5-tert-Butyl-2-methoxybenzenesulfonyl chloride
5-tert-Butyl-2-methoxybenzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₁H₁₅ClO₃S
MDL Number: MFCD08457535
CAS Number: 88041-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR