2,4-Dichloro-5-nitropyrimidine ≥97% for synthesis

Supplier: Matrix Scientific
Warning

021422-25G 021422-100G
101812-056EA 72.31 USD
101812-056 101812-058
2,4-Dichloro-5-nitropyrimidine ≥97% for synthesis
2,4-Dichloro-5-nitropyrimidine

Hazard code(s): Irritant, Lachrymator, Avoid Skin Contact, Store in Freezer

Formula: C₄HCl₂N₃O₂
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1.74 g/cm³ (20 °C)
MDL Number: MFCD00127867
CAS Number: 49845-33-2

Order Now

Specification Test Results

Melting point 28-32°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR