5-Chloronicotinic acid ≥95% for synthesis

Supplier: Matrix Scientific

021377-1G 021377-5G
101811-906EA 38.46 USD
101811-906 101811-908
5-Chloronicotinic acid ≥95% for synthesis
5-Chloronicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄ClNO₂
MW: 157.56
MDL Number: MFCD00128901
CAS Number: 22620-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR