5-Chlorovaleronitrile ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 5-Chloropentanenitrile

021046-25G 021046-100G
101811-260EA 27.69 USD
101811-260 101811-262
5-Chlorovaleronitrile ≥98% for synthesis
5-Chlorovaleronitrile

Hazard code(s): Toxic

Formula: C₅H₈ClN
Boiling Pt: 92…93 °C (10mm)
Density: 1.447 g/cm³
MDL Number: MFCD00001977
CAS Number: 6280-87-1
EINECS: 228-481-4
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 92-93°C/10m
Density 1.447
ND20 1.446


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR