6-Chloro-7-deazapurine ≥98%

Supplier: Matrix Scientific

020966-5G
101811-124EA 63.14 USD
101811-124
6-Chloro-7-deazapurine ≥98%
6-Chloro-7-deazapurine

Hazard: Irritant

Formula: C₆H₄ClN₃
Melting Pt: 183…184 °C
MDL Number: MFCD01686865
CAS Number: 3680-69-1

Order Now

Specification Test Results

Melting point 183-184°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR