[5-(3-Methoxyphenyl)tetrazol-2-yl]acetic acid

Supplier: Matrix Scientific

020749-500MG
101810-704EA 280 USD
101810-704
[5-(3-Methoxyphenyl)tetrazol-2-yl]acetic acid
[5-(3-Methoxyphenyl)tetrazol-2-yl]acetic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₄O₃
MDL Number: MFCD03047607
CAS Number: 436091-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR