3-(Thiophen-2-ylmethoxy)benzaldehyde

Supplier: Matrix Scientific

020484-500MG
101810-108EA 261.54 USD
101810-108
3-(Thiophen-2-ylmethoxy)benzaldehyde
3-(Thiophen-2-ylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₀O₂S
MDL Number: MFCD07801183
CAS Number: 878433-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR