2-Bromo-6-fluoropyridine ≥98%
Supplier: Matrix Scientific
020178-5G
020178-25G
020178-100G
101809-536EA
56.92
USD
101809-536
101809-538
101809-540
2-Bromo-6-fluoropyridine ≥98%
2-Bromo-6-fluoropyridine
Hazard code(s): Toxic
Formula:
C₅H₃BrFN Boiling Pt: 162…164 °C Melting Pt: 30…32 °C |
MDL Number:
MFCD03095258 CAS Number: 144100-07-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Boiling point | 162-164°C |
Melting point | 30-32°C |
ND20 | 1.532 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...