2-Bromo-6-fluoropyridine ≥98%

Supplier: Matrix Scientific

020178-5G 020178-25G 020178-100G
101809-536EA 56.92 USD
101809-536 101809-538 101809-540
2-Bromo-6-fluoropyridine ≥98%
2-Bromo-6-fluoropyridine

Hazard code(s): Toxic

Formula: C₅H₃BrFN
Boiling Pt: 162…164 °C
Melting Pt: 30…32 °C
MDL Number: MFCD03095258
CAS Number: 144100-07-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 162-164°C
Melting point 30-32°C
ND20 1.532


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR