(6-Fluoro-1,2,3,4-tetrahydro-carbazol-9-yl)acetic acid

Supplier: Matrix Scientific

019907-500MG
101809-024EA 280 USD
101809-024
(6-Fluoro-1,2,3,4-tetrahydro-carbazol-9-yl)acetic acid
(6-Fluoro-1,2,3,4-tetrahydro-carbazol-9-yl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₄FNO₂
MDL Number: MFCD07186561
CAS Number: 883541-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR