3-Allylsalicylaldehyde

Supplier: Matrix Scientific

Synonyms: 3-AllylSalicylaldehyde, 3-Allyl-2-hydroxybenzaldehyde

018548-500MG
101806-412EA 63.08 USD
101806-412
3-Allylsalicylaldehyde
3-Allylsalicylaldehyde

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00037364
CAS Number: 24019-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR