2-Fluoro-3-nitropyridine ≥98% for synthesis

Supplier: Matrix Scientific

018339-5G
101805-970EA 60 USD
101805-970
2-Fluoro-3-nitropyridine ≥98% for synthesis
2-Fluoro-3-nitropyridine

Hazard code(s): Toxic

Formula: C₅H₃FN₂O₂
MW: 142.09
MDL Number: MFCD03095068
CAS Number: 1480-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR