7-Bromoindole ≥95%

Supplier: Matrix Scientific
Warning

018335-1G
101805-956EA 18.46 USD
101805-956
7-Bromoindole ≥95%
7-Bromoindole

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₆BrN
Boiling Pt: 85…86 °C (0.2mm)
Melting Pt: 42…44 °C
MDL Number: MFCD00799492
CAS Number: 51417-51-7

Order Now

Specification Test Results

Boiling point 85-86°C/0.2
Melting point 42-44°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR