2,3,4-Trifluorophenylboronic acid for synthesis

Supplier: Matrix Scientific

Synonyms: 2,3,4-Trifluorobenzeneboronic acid, Dihydroxy(2,3,4-trifluorophenyl)borane, (2,3,4-Trifluorophenyl)metaboric acid

018334-5G
101805-952EA 35.38 USD
101805-952
2,3,4-Trifluorophenylboronic acid for synthesis
2,3,4-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MDL Number: MFCD01863168
CAS Number: 226396-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR