2-Fluoro-5-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

018281-25G
101805-810EA 67.69 USD
101805-810
2-Fluoro-5-(trifluoromethyl)pyridine ≥97%
2-Fluoro-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₃F₄N
MDL Number: MFCD07437945
CAS Number: 69045-82-5
EINECS: 400-290-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR