2-Hydroxynicotinic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Hydroxypyridine-3-carboxylic acid, 1,2-Dihydro-2-oxonicotinic acid, 2-Hydroxy-3-pyridinecarboxylic acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic acid

018261-100G
101805-762EA 75.38 USD
101805-762
2-Hydroxynicotinic acid ≥98% for synthesis
2-Hydroxynicotinic acid

Hazard code(s): Irritant

Formula: C₆H₅NO₃
Melting Pt: 258…260 °C
MDL Number: MFCD00010100
CAS Number: 609-71-2
EINECS: 210-198-2

Order Now

Specification Test Results

Melting point 258-260°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR