N-(3-Pyridyl)pivalamide

Supplier: Matrix Scientific

018143-1G
101805-500EA 15.38 USD
101805-500
N-(3-Pyridyl)pivalamide
N-(3-Pyridyl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MDL Number: MFCD00996243
CAS Number: 70298-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR