3-Amino-3-(4-isopropylphenyl)propionic acid

Supplier: Matrix Scientific

016489-500MG
101804-826EA 283.08 USD
101804-826
3-Amino-3-(4-isopropylphenyl)propionic acid
3-Amino-3-(4-isopropylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MW: 207.2700 g/mol
MDL Number: MFCD02663184
CAS Number: 117391-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR