Pyridazine-4,5-dicarboxylic acid ≥98%

Supplier: Matrix Scientific

016114-500MG
101804-128EA 84.62 USD
101804-128
Pyridazine-4,5-dicarboxylic acid ≥98%
Pyridazine-4,5-dicarboxylic acid

Hazard: Irritant

Formula: C₆H₄N₂O₄
Melting Pt: >210 °C (dec.)
MDL Number: MFCD06742784
CAS Number: 59648-14-5

Order Now

Specification Test Results

Melting point 210°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR