4-Bromo-m-cresol ≥95%

Supplier: Matrix Scientific
Warning

016032-5G 016032-25G 016032-100G
101803-940EA 15.38 USD
101803-940 101803-942 101803-944
4-Bromo-m-cresol ≥95%
4-Bromo-m-cresol

Hazard code(s): Irritant

Formula: C₇H₇BrO
Boiling Pt: 142…145 °C (23mm)
Melting Pt: 59…61 °C
MDL Number: MFCD00079723
CAS Number: 14472-14-1
EINECS: 238-464-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR