4-(Cyclohexylmethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

014442-500MG
101803-750EA 278.46 USD
101803-750
4-(Cyclohexylmethoxy)piperidine hydrochloride
4-(Cyclohexylmethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO
MDL Number: MFCD12026488
CAS Number: 1185298-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR