4-[(3-Fluorophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

013667-500MG
101803-652EA 278.46 USD
101803-652
4-[(3-Fluorophenoxy)methyl]piperidine hydrochloride
4-[(3-Fluorophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClFNO
MDL Number: MFCD08445759
CAS Number: 614731-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR