2-Mercapto-4,5,6-trimethylnicotinonitrile ≥98%

Supplier: Matrix Scientific

012601-1G
101802-954EA 144.62 USD
101802-954
2-Mercapto-4,5,6-trimethylnicotinonitrile ≥98%
2-Mercapto-4,5,6-trimethylnicotinonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂S
Melting Pt: 276 °C
MDL Number: MFCD00052657
CAS Number: 128917-84-0

Order Now

Specification Test Results

Melting point 276°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR