2,6-Dichloropyridine-4-methanol ≥98%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine

012589-1G
101802-928EA 83.08 USD
101802-928
2,6-Dichloropyridine-4-methanol ≥98%
2,6-Dichloropyridine-4-methanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
Melting Pt: 127…130 °C
MDL Number: MFCD00052638
CAS Number: 101990-69-6

Order Now

Specification Test Results

Melting point 127-130°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR