4-(Methylsulfonyl) phenylacetonitrile ≥98%

Supplier: Matrix Scientific

012551-1G
101802-834EA 109.23 USD
101802-834
4-(Methylsulfonyl) phenylacetonitrile ≥98%
4-(Methylsulfonyl) phenylacetonitrile

Hazard: Irritant

Formula: C₉H₉NO₂S
Melting Pt: 122…124 °C
MDL Number: MFCD00079775
CAS Number: 25025-07-4

Order Now

Specification Test Results

Melting point 122-124°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR