5-Methyl-2-thenoic acid ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
5-Methylthiophene-2-carboxylic acid
012537-5G
012537-25G
101802-796EA
15.38
USD
101802-796
101802-798
5-Methyl-2-thenoic acid ≥98% for synthesis
5-Methyl-2-thenoic acid
Hazard code(s): Irritant
Formula:
C₆H₆O₂S Melting Pt: 136…138 °C |
MDL Number:
MFCD00005439 CAS Number: 1918-79-2 EINECS: 217-640-3 |
Specification Test Results
Melting point | 136-138°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...