3-Fluoro-p-anisaldehyde ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methoxybenzaldehyde, 3-Fluoro-4-anisaldehyde

011203-5G 011203-25G
101800-118EA 61.54 USD
101800-118 101800-120
3-Fluoro-p-anisaldehyde ≥98% for synthesis
3-Fluoro-p-anisaldehyde

Hazard code(s): Irritant, Air Sensitive, Refrigerate

Formula: C₈H₇FO₂
MW: 154.14
Melting Pt: 34…35 °C
MDL Number: MFCD00003349
CAS Number: 351-54-2
EINECS: 206-514-3

Order Now

Specification Test Results

Melting point 34-35°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR