2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide

Supplier: Matrix Scientific

Synonyms: 2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

010726-500MG
101799-088EA 84.62 USD
101799-088
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide

Hazard: Irritant

Formula: C₈H₁₀N₂OS
MW: 182.25 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00122463
CAS Number: 77651-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR