2-Azepan-1-ylethylamine

Supplier: Matrix Scientific

010564-500MG
101798-768EA 207.69 USD
101798-768
2-Azepan-1-ylethylamine
2-Azepan-1-ylethylamine

Hazard: Irritant

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Boiling Pt: 212 °C
Melting Pt: 200 °C
MDL Number: MFCD00046120
CAS Number: 51388-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR