5-(4-Fluorophenyl)[1,3,4]thiadiazol-2-ylamine

Supplier: Matrix Scientific

Synonyms: 5-(4-Fluorophenyl)-1,3,4-thiadiazol-2-amine

009949-500MG
101797-572EA 267.69 USD
101797-572
5-(4-Fluorophenyl)[1,3,4]thiadiazol-2-ylamine
5-(4-Fluorophenyl)[1,3,4]thiadiazol-2-ylamine

Hazard: Irritant

Formula: C₈H₆FN₃S
Melting Pt: 211 °C
MDL Number: MFCD00981219
CAS Number: 942-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR