Cyclohexylboronic acid pinacol ester ≥96%

Supplier: Matrix Scientific

Synonyms: Cyclohexylboronic acid Pinacol ester, 2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohexane

009774-1G
101797-232EA 109.23 USD
101797-232
Cyclohexylboronic acid pinacol ester ≥96%
Cyclohexylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₂H₂₃BO₂
MDL Number: MFCD04038749
CAS Number: 87100-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR