4-Carboxyphenylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

009765-1G 009765-5G 009765-25G
101797-200EA 15.38 USD
101797-200 101797-202 101797-204
4-Carboxyphenylboronic acid pinacol ester ≥97%
4-Carboxyphenylboronic acid pinacol ester

Hazard: Irritant/Refrigerate

Formula: C₁₃H₁₇BO₄
Melting Pt: 227…231 °C
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR