Nonafluoropentanal hydrate

Supplier: Matrix Scientific

009426-5G
101797-070EA 135.38 USD
101797-070
Nonafluoropentanal hydrate
Nonafluoropentanal hydrate

Hazard: Irritant

Formula: C₅H₃F₉O₂
MDL Number: MFCD04115826
CAS Number: 907607-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR