4-(Methylsulfonyl)-2-nitrobenzaldehyde

Supplier: Matrix Scientific

009325-1G
101796-868EA 244.62 USD
101796-868
4-(Methylsulfonyl)-2-nitrobenzaldehyde
4-(Methylsulfonyl)-2-nitrobenzaldehyde

Hazard: Irritant

Formula: C₈H₇NO₅S
MDL Number: MFCD04037929
CAS Number: 849035-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR