Methyl perfluoro-3,6-dioxaheptanoate ≥98%

Supplier: Matrix Scientific

009034-5G 009034-25G
101796-164EA 118.46 USD
101796-164 101796-166
Methyl perfluoro-3,6-dioxaheptanoate ≥98%
Methyl perfluoro-3,6-dioxaheptanoate

Hazard: Irritant

Formula: C₆H₃F₉O₄
MDL Number: MFCD03094140
CAS Number: 39187-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR