4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ethyl ester

Supplier: Matrix Scientific

Synonyms: Ethyl 3,5-dimethyl-4-formyl-1H-pyrrole-2-carboxylate

008368-500MG
101794-544EA 230.77 USD
101794-544
4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ethyl ester
4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
Melting Pt: 145 °C
MDL Number: MFCD00030384
CAS Number: 2199-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR