5-Amino-2-(p-tolyl)-2H-benzotriazole

Supplier: Matrix Scientific

007970-500MG
101793-756EA 258.46 USD
101793-756
5-Amino-2-(p-tolyl)-2H-benzotriazole
5-Amino-2-(p-tolyl)-2H-benzotriazole

Hazard: Irritant

Formula: C₁₃H₁₂N₄
MDL Number: MFCD00477196
CAS Number: 6659-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR