2-Methoxy-5-methylphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-5-methylbenzeneboronic acid

007411-1G
101792-692EA 156.92 USD
101792-692
2-Methoxy-5-methylphenylboronic acid ≥97%
2-Methoxy-5-methylphenylboronic acid

Hazard: Irritant

Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
Melting Pt: 80…84 °C
MDL Number: MFCD02179465
CAS Number: 127972-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR