1,3-Dichloro-1,1-difluoropropane ≥98%

Supplier: Matrix Scientific

007194-5G 007194-25G 007194-100G
101792-156EA 63.08 USD
101792-156 101792-158 101792-160
1,3-Dichloro-1,1-difluoropropane ≥98%
1,3-Dichloro-1,1-difluoropropane

TCA listed: F
Hazard: Ozone Depleter

Formula: C₃H₄Cl₂F₂
Boiling Pt: 81…81.5 °C
Density: 1.339
MDL Number: MFCD01320726
CAS Number: 819-00-1

Order Now

Specification Test Results

Boiling point 81-81.5°C
Density 1.339


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR