7-Hydroxy-4-(trifluoromethyl)coumarin ≥98%

Supplier: Matrix Scientific

007028-5G
101791-866EA 113.85 USD
101791-866
7-Hydroxy-4-(trifluoromethyl)coumarin ≥98%
7-Hydroxy-4-(trifluoromethyl)coumarin

Hazard: Irritant

Formula: C₁₀H₅F₃O₃
Melting Pt: 178…180 °C
MDL Number: MFCD00037578
CAS Number: 575-03-1

Order Now

Specification Test Results

Melting point 178-180°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR